EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Alkynyliodonium salts in organic synthesis. Application to the total synthesis of the tropoloisoquinoline alkaloid pareitropone.
PMID 12296717
Abstract
The synthesis of the tropoloisoquinoline alkaloid pareitropone has been accomplished in 14 steps from 2,3,4-trimethoxybenzoic acid. The key transformations include the generation of an alkylidenecarbene intermediate through intramolecular addition of a tosylamide anion to an alkynyliodonium salt, and the cycloaddition of that carbene to a peri positioned aromatic ring to afford a cycloheptatrienylidene product featuring the intact pareitropone skeleton.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
189790
2,3,4-Trimethoxybenzoic acid, ≥98%
C10H12O5