EMAIL THIS PAGE TO A FRIEND
Clinical and experimental immunology
Elevated levels of soluble CD163 in sera and fluids from rheumatoid arthritis patients and inhibition of the shedding of CD163 by TIMP-3.
PMID 12296867
Abstract
The aim of the present study was to evaluate levels of soluble CD 163 in sera and fluids from rheumatoid arthritis (RA) patients and elucidate the mechanism that regulates the shedding of CD163. Levels of soluble CD163 in sera and fluids from RA patients were examined by a sandwich enzyme immunoassay and Western blotting. To determine the effects of tissue inhibitors of metalloproteinase (TIMPs) on the shedding of CD163 from monocytes/macrophages, levels of soluble CD163 in cultures of monocytes/macrophages and the expression of CD163 on monocytes/macrophages in the presence or absence of TIMPs were examined by a sandwich enzyme immunoassay and flow cytometry, respectively. The clinical marker that was most associated with serum levels of soluble CD163 was levels of CRP. TIMP-3, but not TIMP-1 or TIMP-2, inhibited the shedding of CD163 from monocytes/macrophages. It was shown that serum levels of soluble CD163 are a sensitive and reliable marker to monitor activated macrophages in synovitis from RA patients and the results imply that the responsible proteinase for the shedding of CD163 is not a member of the matrix metalloproteinases, but is likely to be a member of ADAMs.