EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Palladium-catalyzed asymmetric allylic alkylation of alpha-aryl ketones.
PMID 12298076
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T19208
β-Tetralone, 98%
C10H10O