EMAIL THIS PAGE TO A FRIEND
Organic letters
A general, efficient, and inexpensive catalyst system for the coupling of aryl iodides and thiols.
PMID 12323058
Abstract
An efficient copper-catalyzed carbon-sulfur bond formation reaction was developed. This method is particularly noteworthy given its experimental simplicity, high generality, and exceptional level of functional group toleration and the low cost of the catalyst system. [reaction: see text]
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
392510
1-Iodo-3,5-dimethylbenzene, 99%
C8H9I