EMAIL THIS PAGE TO A FRIEND
Zhurnal evoliutsionnoi biokhimii i fiziologii
[Phosphororganic compound O,O-dimethyl-O-(2,2-dichlorovinyl)phosphate as a selective inhibitor for separate analysis of the roach Rutilus rutilus acetylcholinesterase and butylcholinesterase activities ].
PMID 12325258
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T1505
Tetraisopropyl pyrophosphoramide, butyrylcholinesterase inhibitor
C12H32N4O3P2