EMAIL THIS PAGE TO A FRIEND
British journal of sports medicine
Urine nandrolone metabolites: false positive doping test?
PMID 12351328
Abstract
The aim of this review is to analyse the studies on nandrolone metabolism to determine if it is possible for an athlete to test positive for nandrolone without having ingested or injected nandrolone.