EMAIL THIS PAGE TO A FRIEND
Inorganic chemistry
Simultaneous N7,O6-binding of guanine to two zinc centers and its possible biological significance.
PMID 12354023
Abstract
The reaction of ZnCl(2) with 9-ethylguanine (9-EtGH) produced a novel dinuclear Zn(II) complex, [Zn(2)Cl(4)(H(2)O)(mu-9-EtGH-N7,O6)(9-EtGH-N7), 1. The X-ray structure analysis (monoclinic, P2(1) (No. 4), a = 11.0636(6) A, b = 6.6546(4) A, c = 15.9630(9) A, beta = 101.069(1) degrees, V = 1153.4(1) A(3), Z = 2) revealed that one of the tetrahedrally coordinated Zn(II) atoms binds to the N7 site of 9-EtGH and to the exocyclic O6 atom of another 9-EtGH molecule. The remaining Zn(II) atom binds to the N7 site of the second 9-EtGH moiety.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E4267
9-Ethylguanine
C7H9N5O