EMAIL THIS PAGE TO A FRIEND
Methods in molecular biology (Clifton, N.J.)
Rapid liquid-phase combinatorial synthesis of heterocyclic libraries.
PMID 12357923
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
129550
4-(2-Keto-1-benzimidazolinyl)piperidine, 98%
C12H15N3O