EMAIL THIS PAGE TO A FRIEND
Molecular and cellular neurosciences
Cannabimimetic activity, binding, and degradation of stearoylethanolamide within the mouse central nervous system.
PMID 12359156
Abstract
Stearoylethanolamide (SEA) is present in human, rat, and mouse brain in amounts comparable to those of the endocannabinoid anandamide (arachidonoylethanolamide, AEA). Yet, the biological activity of SEA has never been investigated. We report that SEA has the same effects as AEA on catalepsy, motility, analgesia, and body temperature of mice and that specific binding sites for SEA are present in mouse brain and are most abundant in cortex. Pharmacological experiments and the use of knockout mice demonstrated that these sites are different from cannabinoid receptors, are not coupled to G proteins, and regulate different signaling pathways. Mouse brain has also a specific SEA membrane transporter and a fatty acid amide hydrolase able to cleave SEA, with the same regional distribution as the binding sites of this lipid. Moreover, SEA potentiates the decrease of cAMP induced by AEA in mouse cortical slices, suggesting that SEA might also be an "entourage" compound.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S8439
Stearoyl ethanolamide, ≥98%, crystalline
C20H41NO2