EMAIL THIS PAGE TO A FRIEND
Bulletin of environmental contamination and toxicology
Isotopic characterization of six major brands of white basic lead carbonate paint pigments.
PMID 12375107
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
336378
Lead(II) carbonate, ACS reagent
CO3Pb