EMAIL THIS PAGE TO A FRIEND
Science (New York, N.Y.)
Modulation of acetaminophen-induced hepatotoxicity by the xenobiotic receptor CAR.
PMID 12376703
Abstract
We have identified the xenobiotic receptor CAR (constitutive androstane receptor) as a key regulator of acetaminophen metabolism and hepatotoxicity. Known CAR activators as well as high doses of acetaminophen induced expression of three acetaminophen-metabolizing enzymes in wild-type but not in CAR null mice, and the CAR null mice were resistant to acetaminophen toxicity. Inhibition of CAR activity by administration of the inverse agonist ligand androstanol 1 hour after acetaminophen treatment blocked hepatotoxicity in wild type but not in CAR null mice. These results suggest an innovative therapeutic approach for treating the adverse effects of acetaminophen and potentially other hepatotoxic agents.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A2480
5α-Androstan-3β-ol, powder
C19H32O