EMAIL THIS PAGE TO A FRIEND
Journal of molecular biology
The structure of an FF domain from human HYPA/FBP11.
PMID 12381297
Abstract
The FF domain is a 60 amino acid residue phosphopeptide-binding module found in a variety of eukaryotic proteins including the transcription elongation factor CA150, the splicing factor Prp40 and p190RHOGAP. We have determined the structure of an FF domain from HYPA/FBP11. The domain is composed of three alpha helices arranged in an orthogonal bundle with a 3(10) helix in the loop between the second and third alpha helices. The structure differs from those of other phosphopeptide-binding domains and represents a novel phosphopeptide-binding fold.