EMAIL THIS PAGE TO A FRIEND
Analytical biochemistry
A spectrophotometric assay for feruloyl esterases.
PMID 12381367
Abstract
We have developed a spectrophotometric assay for the quantitative determination of feruloyl esterase activity based on release of 4-nitrophenol from a novel substrate, 4-nitrophenyl ferulate in an emulsion of Triton X-100 in aqueous buffer solution. The release of 4-nitrophenol was linear with reaction time at an early stage of the reaction with various esterase preparations. The method proposed here is accurate, rapid, and easy to perform.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
61716
4-Nitrophenyl dodecanoate, ≥98.0% (GC)
C18H27NO4