EMAIL THIS PAGE TO A FRIEND
Gene
Genetics of isoprenoid biosynthesis in Paracoccus zeaxanthinifaciens.
PMID 12384294
Abstract
The genes coding for all the enzymes involved in the conversion of acetyl-CoA to farnesyl diphosphate (FPP) in the zeaxanthin-producing bacterium Paracoccus zeaxanthinifaciens were cloned and characterized. Two genes encoding enzymes catalysing the condensation of two acetyl-CoA molecules to acetoacetyl-CoA were found. The six enzymes involved in the conversion of acetyl-CoA and acetoacetyl-CoA to isopentenyl diphosphate (IPP) and dimethylallyl diphosphate (DMAPP) are grouped in an operon, designated the mevalonate operon. The gene encoding the enzyme catalysing two consecutive condensations, IPP and DMAPP to geranyl diphosphate (GPP) and IPP and GPP to FPP, is not clustered with any other gene encoding an enzyme of the isoprenoid pathway. Genes encoding enzymes involved in the biosynthesis of poly-hydroxyalkanoate and non-carotenoid isoprenoids found in P. zeaxanthinifaciens are also presented.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A1625
Acetoacetyl coenzyme A sodium salt hydrate, cofactor for acyl transfer
C25H40N7O18P3S · xNa+ · yH2O