EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Solid-state and solution studies of a tetrameric capsule and its guests.
PMID 12386850
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
146048
2-Adamantanone, ReagentPlus®, 99%
C10H14O