EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Characterization of Drosophila carboxypeptidase D.
PMID 12393882
Abstract
Metallocarboxypeptidase D (CPD), is a 180-kDa protein that contains three carboxypeptidase-like domains, a transmembrane domain, and a cytosolic tail and which functions in the processing of proteins that transit the secretory pathway. An initial report on the Drosophila melanogaster silver gene indicated a CPD-like protein with only two and a half carboxypeptidase-like domains with no transmembrane region (Settle, S. H., Jr., Green, M. M., and Burtis, K. C. (1995) Proc. Natl. Acad. Sci. U. S. A. 92, 9470-9474). A variety of bioinformatics and experimental approaches were used to determine that the Drosophila silver gene corresponds to a CPD-like protein with three carboxypeptidase-like domains, a transmembrane domain, and a cytosolic tail. In addition, two alternative exons were found, which result in proteins with different carboxypeptidase-like domains, termed domains 1A and 1B. Northern blot, reverse transcriptase PCR, and sequence analysis were used to confirm the presence of the various mRNA forms. Individual domains of Drosophila CPD were expressed in insect Sf9 cells using the baculovirus expression system. Media from domain 1B- and domain 2-expressing cells showed substantial enzymatic activity, whereas medium from domain 1A-expressing cells was no different from cells infected with wild-type virus. Domains 1B and 2 were purified, and the enzymatic properties were examined. Both enzymes cleaved substrates with C-terminal Arg or Lys, but not Leu, and were inhibited by conventional metallopeptidase inhibitors and some divalent cations. Drosophila domain 1B is more active at neutral pH and greatly prefers C-terminal Arg over Lys, whereas domain 2 is more active at pH 5-6 and slightly prefers C-terminal Lys over Arg. The differences in pH optima and substrate specificity between Drosophila domains 1B and 2 are similar to the differences between duck CPD domains 1 and 2, suggesting that these properties are essential to CPD function.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C6742 Carboxypeptidase W from wheat, ≥50 units/mg protein