EMAIL THIS PAGE TO A FRIEND
The Journal of pharmacy and pharmacology
Endothelium-dependent vasorelaxation induced by L-carnitine in isolated aorta from normotensive and hypertensive rats.
PMID 12396307
Abstract
The aim of this work was to investigate the mechanism of the vasodilatory effect induced by L-carnitine. Relaxation produced by L-carnitine was studied in rat aortic rings with and without functional endothelium, pre-contracted with phenylephrine by adding cumulative doses of L-carnitine (10(-7) to 10(-3) M). The relaxation evoked by L-carnitine reached higher values in aortic rings from spontaneously hypertensive rats than those obtained in arteries from normotensive rats; no relaxation was produced in de-endothelialized arteries. However, in the presence of N(G)-nitro-L-arginine (3 x 10(-5) M, a nitric oxide synthase inhibitor), Ro 68070 (10(-4) M, a thromboxane synthetase inhibitor-thromboxane A2/prostaglandin H2 receptor antagonist) or ICI 192605 (10(-5) M, a thromboxane A2 receptor antagonist) the relaxant response to L-carnitine was significantly inhibited. These results show that L-carnitine induced endothelium-dependent relaxation in the rat aorta and the mechanism of this relaxation appeared to be mostly mediated by endothelial production of nitric oxide but#10; also could involve prevention of the action of cyclooxygenase endothelial products acting on the thromboxane A2/prostaglandin H2 receptor.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I2536
ICI 192605, ≥98% (HPLC)
C22H23ClO5