EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
New supramolecular organization for a glycoluril: chiral hydrogen-bonded ribbons.
PMID 12397991
Abstract
A series of substituted glycoluril molecules exhibits a substantial twist of the fused five-membered rings and assembles exclusively chiral hydrogen-bonded ribbons in the solid-state.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G7305
Glycoluril, 97%
C4H6N4O2