EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences
Stable isotope dilution high-performance liquid chromatography-electrospray ionization mass spectrometry method for endogenous 2- and 4-hydroxyestrones in human urine.
PMID 12401358
Abstract
A sensitive, precise and accurate stable isotope dilution high-performance liquid chromatography-electrospray ionization mass spectrometry method has been developed for measuring endogenous 2- and 4-hydroxyestrones, the main catechol estrogens in human urine. Compared to the published methods using gas chromatography-mass spectrometry, this approach simplifies sample preparation and increases the throughput of analysis. The unique part of our method is the use of a simple and rapid derivatization step that forms a hydrazone at the C-17 carbonyl group of catechol estrogens. This derivatization step has greatly enhanced method sensitivity as well as HPLC separability of 2- and 4-hydroxyestrones. Standard curves were linear over a 100-fold calibration range with correlation coefficients for the linear regression curves typically greater than 0.996. The lower limit of quantitation for each catechol estrogen is 1 ng per 10-ml urine sample, with an accuracy of 97-99% and overall precision, including the hydrolysis, extraction and derivatization steps, of 1-3% for samples prepared concurrently and 2-11% for samples prepared in several batches. This method is adequate for measuring the low endogenous levels of catechol estrogens in urine from postmenopausal women.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H6012
4-Hydroxyestrone, ≥90% (HPLC)
C18H22O3