EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
About the Antiaromaticity of Planar Cyclooctatetraene I thank Jens Panitzky for the additional DFT calculations, and Professor Jay Siegel, La Jolla, CA, and Professor Otto Ermer, Cologne, for helpful comments.
PMID 12404467
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
138924
Cyclooctatetraene, 98%
C8H8