EMAIL THIS PAGE TO A FRIEND
Journal of agricultural and food chemistry
Development of an enzyme-linked immunosorbent assay for the organophosphorus insecticide bromophos-ethyl.
PMID 12405760
Abstract
A competitive enzyme-linked immunosorbent assay (ELISA) was developed for the quantitative detection of the organophosphorus insecticide bromophos-ethyl. Four bromophos-ethyl derivatives (haptens) were synthesized and were coupled to carrier proteins through the pesticide thiophosphate group to use as an immunogen or as a coating antigen. Rabbits were immunized with either one of two haptens coupled to bovine serum albumin for production of polyclonal antibodies, and the sera were screened against one of the haptens coupled to ovalbumin. Using the serum with highest titer, an antigen-coated ELISA was developed, which showed an IC(50) of 3.9 ng/mL with a detection limit of 0.3 ng/mL (20% inhibition). An antibody-coated ELISA using an enzyme tracer was also developed, which showed an IC(50) of 6.5 ng/mL with a detection limit of 1.0 ng/mL (20% inhibition). The antibodies showed negligible cross-reactivity with other organophosphorus pesticides except with the insecticides bromophos-methyl and chlorpyrifos in the antibody-coated assay only. Recoveries of bromophos-ethyl from fortified crop and water samples ranged from 82 to 128% and from 95 to 127%, respectively.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
33399
Bromophos-ethyl, PESTANAL®, analytical standard
C10H12BrCl2O3PS
33400
Bromophos-methyl, PESTANAL®, analytical standard
C8H8BrCl2O3PS