EMAIL THIS PAGE TO A FRIEND
Nature biotechnology
Neural stem cells, scaffolds, and chaperones.
PMID 12410250
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M103
1-Methyl-4-(2′-methylphenyl)-1,2,3,6-tetrahydropyridine hydrochloride, solid
C13H17N · HCl