EMAIL THIS PAGE TO A FRIEND
FEBS letters
Mechanisms for 2-methoxyestradiol-induced apoptosis of prostate cancer cells.
PMID 12417302
Abstract
Prostate and breast carcinomas are sex hormone-related carcinomas, which are known to be associated with an over-expression of the proto-oncogene Bcl-2. Here, we report that 2-methoxyestradiol (2-ME), an endogenous metabolite of estrogen that does not bind to nuclear estrogen receptors, effectively induces apoptosis in Bcl-2-expressing human prostate and breast carcinoma cells in vitro and in a rat prostate tumor model in vivo. In several cell lines derived from prostate, breast, liver and colorectal carcinomas, 2-ME treatment led to an activation of c-Jun N-terminal kinase (JNK) and phosphorylation of Bcl-2, which preceded the induction of apoptosis. In summary, our data suggest that 2-ME induces apoptosis in epithelial carcinomas by causing phosphorylation of JNK, which appears to be correlated with phosphorylation of Bcl-2.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M6383
2-Methoxyestradiol, powder
C19H26O3