EMAIL THIS PAGE TO A FRIEND
Methods in enzymology
Detection of covalent tetrahedral adducts by differential isotope shift 13C NMR: acetyl-enzyme reaction intermediate formed by 3-hydroxy-3-methylglutaryl-CoA synthase.
PMID 12418229
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A1625
Acetoacetyl coenzyme A sodium salt hydrate, cofactor for acyl transfer
C25H40N7O18P3S · xNa+ · yH2O