EMAIL THIS PAGE TO A FRIEND
Respiratory medicine
Efficacy and safety of oxitropium bromide, theophylline and their combination in COPD patients: a double-blind, randomized, multicentre study (BREATH Trial).
PMID 12418585
Abstract
We compare the efficacy including spirometry, peak expiratory flow (PEFR) and quality of life and safety of an 8-week treatment with inhaled oxitropium, theophylline or their combination in patients with mild-to-severe chronic obstructive pulmonary disease (COPD). We conducted a multicentre, double-blind, double-dummy randomized, parallel-group study at 29 Italian outpatients clinics. A group of 236 patients with mild-to-severe COPD (baseline FEV1 < or = 70% of predicted value) were recruited. Treatments were as follows: Inhaled oxitropium bromide 200 microg (N=75), sustained-release oral theophylline 300 mg (N=81) or their combination (N=80), taken twice daily. Spirometry (FEV1 and FVC) was evaluated every 4 weeks, and morning and evening PEFR (before and 2-4 h after drug intake) was measured daily. Symptoms, cough and dysponea, were recorded daily. Health status was evaluated at baseline and week 8 using the disease specific St George' Respiratory Questionnaire (SGRQ). Any adverse event occurring during the treatment period was recorded on a diary card. FEV1 and FVC improved in all the groups at 4 and 8 weeks, but the difference between treatment groups did not reach statistically significant levels. Differences between groups in pre-dosing morning and evening PEFR were not significant. Post-dosing morning and evening PEFR were increased and the largest increase was seen in patients treated with both drugs. However, differences between groups was significant only for evening values (P=0.008). The proportion of patients who experienced a decrease in symptoms was high in all groups but no differences among groups were observed. SGRQ total scores decreased in all treatment groups after 8 weeks, particularly in the oxitropium and combination groups. Clinically significant change (> or = 4 units) was only observed in patients treated with oxitropium bromide whether with or without theophylline. Adverse events related to treatments were higher in the group treated with theophylline alone (P < 0.02). We conclude that inhaled oxitropium bromide alone was associated with an improvement in FEV1, PEFR and symptoms in patients with COPD that was not statistically different from that of oral theophylline alone or of the combination of both drugs. Oxitropium bromide in combination with theophylline provided a greater improvement in evening post-dosing PEFR. Oxitropium bromide alone or in combination with theophylline improved the quality of life better than theophylline alone.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Y0000709 Oxitropium bromide, European Pharmacopoeia (EP) Reference Standard
C19H26BrNO4