EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Chemical modification of reveromycin A and its biological activities.
PMID 12419362
Abstract
Various derivatives of reveromycin A, a novel inhibitor of eukaryotic cell growth, were prepared and their inhibitory effects on both isoleucyl-tRNA synthetase activity and in vitro protein synthesis, and activities on the morphological reversion of src(ts)-NRK cells were assayed. The C5 hydroxyl group and C24 carboxyl group are particularly important for these activities.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
R0654
Reveromycin A, ≥98% (HPLC), solid
C36H52O11