EMAIL THIS PAGE TO A FRIEND
Farmaco (Societa chimica italiana : 1989)
Synthesis and antitumor activity of 1,3-benzodioxole derivatives.
PMID 12420881
Abstract
A series of 1,3-benzodioxoles (5-19) was synthesized and evaluated for their in vitro antitumor activity against human tumor cell lines. Some derivatives exhibited tumor growth inhibition activity. In particular, 6-(4-aminobenzoyl)-1,3-benzodioxole-5-acetic acid methyl ester 8, the most active compound of the series, possesses a significant growth inhibitory activity on 52 cell lines at concentrations ranging from 10(-7) to 10(-5) M.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
159166
1,3-Benzodioxole, 99%
C7H6O2