EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Modification of the upper rim of homooxacalix[3]arenes and complexation between a nitrohomooxacalix[3]arene derivative and n-hexylamine.
PMID 12423145
Abstract
Several functional groups were introduced on the upper rim of (lower rim free) homooxacalix[3]arene for the first time. The swinging nitrohomooxacalix[3]arene host 1 was fixed in the cone conformation by complexation with n-hexylamine.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
53131
Hexylamine, purum, ≥98.0% (GC)
C6H15N
219703
Hexylamine, 99%
C6H15N