EMAIL THIS PAGE TO A FRIEND
Bollettino chimico farmaceutico
Spectrophotometric determination of nikethamide by charge--transfer complexation with chloranilic acid.
PMID 12426818
Abstract
A Spectrophotometric method is described for assay of nikethamide which was based on charge transfer complexation between it and chloranilic acid. They form a complex with stoichiometric ratio of 1:1 and showed l-max at 535 nm. Maximum complexation occurred 30 min after mixing the reactants and was stable for 12 hr. The molar absorptivity, association constant and free energy for the complex were 1.05 x 1.0(2), 4.40 x 10(-2) litre mole-1 and 1819.02 kcal respectively. Conformity to Beer's Law was within the concentration range of 1-2.5 x 10(-4), which enabled the assay of dosage forms of the drug at microquantities.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
72372
N,N-Diethylnicotinamide, tested according to Ph.Eur.
C10H14N2O
D98807
N,N-Diethylnicotinamide, 99%
C10H14N2O