EMAIL THIS PAGE TO A FRIEND
Proceedings of the Western Pharmacology Society
Efficacy and tolerability of acemetacin, a non-steroidal anti-inflammatory drug, in Mexican patients: result of the ETAPAM Study.
PMID 12434547
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A1674
Acemetacin, analytical standard
C21H18ClNO6