EMAIL THIS PAGE TO A FRIEND
FEBS letters
New potent inhibitors of aquaporins: silver and gold compounds inhibit aquaporins of plant and human origin.
PMID 12435590
Abstract
Silver and gold compounds were tested as potential inhibitors of aquaporins of plant- and human origin. Silver as AgNO(3) or silver sulfadiazine inhibited with high potency (EC(50) 1-10 microM) the water permeability of the peribacteroid membrane from soybean (containing Nodulin 26), the water permeability of plasma membrane from roots (containing plasma membrane integral proteins), and the water permeability of human red cells (containing aquaporin 1). Gold as HAuCl(4) was less effective but still inhibited peribacteroid membrane water permeability (EC(50)=10 microM). Silver and gold are more potent inhibitors of aquaporins than the presently widely used mercury containing compounds.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
230405
Mercury(I) chloride, ACS reagent, ≥99.5%
Cl2Hg2