EMAIL THIS PAGE TO A FRIEND
Zeitschrift fur Naturforschung. C, Journal of biosciences
Chloroacetamides affect the plasma membrane.
PMID 12440722
Abstract
In the present study membrane fatty acids were analyzed to find a link between the biosynthesis inhibition of very-long-chain fatty acids and the phytotoxic effects of herbicidal chloroacetamides. Accordingly, we have isolated membranes of cucumber seedlings (Cucumis sativus) by two-phase partitioning and analyzed their fatty acid content. Saturated VLCFAs ranging from C20 to C26 were found in high amounts (22%) in the plasma membrane fraction. Non-modified VLCFAs were predominantly present in phospholipids, while saturated 2-hydroxylated VLCFAs were identified in cerebrosides. Treatment of intact seedlings with chloroacetamides markedly reduced the VLCFA content in the plasma membrane. This result could be specified by fatty-acid labeling using [14C]malonate as a substrate for fatty acid elongation. De novo incorporation of VLCFAs into the plasma membrane and into microsomal membranes, respectively, was severely impaired by chloroacetamides with I50 values between 10 to 100 nM. These results confirm the previous finding that chloroacetamides inhibit VLCFA biosynthesis localized in the microsomes (Böger et al., Pest Manage. Sci. 56, 497-508, 2000). The direct consequence of this inhibition is a strong decrease of VLCFAs required as constituents of the plasma membrane and the substitution by shorter acyl chains. Apparently, physical properties and function of the plasma membrane are affected eventually leading to death of the plant.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C0267
2-Chloroacetamide, ≥98%
C2H4ClNO
22790
2-Chloroacetamide, ≥98.0% (HPLC)
C2H4ClNO