EMAIL THIS PAGE TO A FRIEND
Annals of neurology
Coenzyme Q-responsive Leigh's encephalopathy in two sisters.
PMID 12447928
Abstract
A 31-year-old woman had encephalopathy, growth retardation, infantilism, ataxia, deafness, lactic acidosis, and increased signals of caudate and putamen on brain magnetic resonance imaging. Muscle biochemistry showed succinate:cytochrome c oxidoreductase (complex II-III) deficiency. Both clinical and biochemical abnormalities improved remarkably with coenzyme Q10 supplementation. Clinically, when taking 300mg coenzyme Q10 per day, she resumed walking, gained weight, underwent puberty, and grew 20cm between 24 and 29 years of age. Coenzyme Q10 was markedly decreased in cerebrospinal fluid, muscle, lymphoblasts, and fibroblasts, suggesting the diagnosis of primary coenzyme Q10 deficiency. An older sister has similar clinical course and biochemical abnormalities. These findings suggest that coenzyme Q10 deficiency can present as adult Leigh's syndrome.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C9538
Coenzyme Q10, ≥98% (HPLC)
C59H90O4