EMAIL THIS PAGE TO A FRIEND
Phytochemistry
Organ-specific analysis of phenylphenalenone-related compounds in Xiphidium caeruleum.
PMID 12453574
Abstract
The distribution pattern of phenylphenalenone-type compounds was investigated in vegetative and reproductive organs of Xiphidium caeruleum. The highest total molar concentration, up to 30 micromol g(-1) fr. wt, was detected in the root tip and the stamen. Accumulation of specific phenylphenalenone-related metabolites including glycosides was found in the hypogeal plant parts, the leaves, and the reproductive organs of the inflorescence. Putative biosynthetic relationships and the role of these compounds in plant defence are discussed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P10801
Perinaphthenone, 97%
C13H8O