EMAIL THIS PAGE TO A FRIEND
FEBS letters
A large number of novel Ergtoxin-like genes and ERG K+-channels blocking peptides from scorpions of the genus Centruroides.
PMID 12459475
Abstract
Twenty-three novel sequences similar to Ergtoxin (ErgTx) were obtained by direct sequencing of peptides or deduced from gene cloned using cDNAs of venomous glands of Centruroides (C.) elegans, C. exilicauda, C. gracilis, C. limpidus limpidus, C. noxius and C. sculpturatus. These peptides have from 42 to 47 amino acid residues cross-linked by four disulfide bridges. They share sequence similarities (60-98% compared with ErgTx1) and were shown to block ERG K(+)-channels of F-11 clone (N18TG-2xrat DRG) cultured cells. An unrooted phylogenetic tree analysis of these peptides showed that they conform at least five different subfamilies, of which three are novel subfamilies.