EMAIL THIS PAGE TO A FRIEND
Postgraduate medicine
Potential agents of chemical warfare. Worst-case scenario protection and decontamination methods.
PMID 12462190
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
23261
Trichloromethyl chloroformate, ≥97.0% (GC)
C2Cl4O2