EMAIL THIS PAGE TO A FRIEND
Nutrition reviews
The regulation of the thyroid-stimulating hormone of the anterior pituitary gland by thyroid hormone and by 9-cis-retinoic acid.
PMID 12462521
Abstract
Thyroid-stimulating hormone, TSH, of the anterior pituitary gland is regulated by the binding of the thyroid hormone-activated thyroid receptor to the TSH gene at the same time as the binding of the 9-cis-retinoic acid-activated retinoid X receptor to the same gene. Both interactions, separately or simultaneously, can suppress and thus regulate the expression of the TSH gene.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
R4643
9-cis-Retinoic acid, ≥98% (HPLC)
C20H28O2