EMAIL THIS PAGE TO A FRIEND
Solid state nuclear magnetic resonance
A study of a moleculartweezer host-guest system by a combination of quantum-chemical calculations and solid-state NMR experiments.
PMID 12469808
Abstract
A study of a host-guest system consisting of a naphthalene-spaced tweezer with a 1,4 dicyanobenzene guest molecule is presented. The complex is investigated using a combination of quantum-chemical calculations and solid-state NMR experiments. The advantages of such an approach are illustrated. The focus is on the calculation of (1) 1H NMR and (2) 13C NMR chemical shifts for model fragments of the solid-state structure, (3) the analysis of host-guest interactions important for molecular recognition, and (4) the investigation of the process of a guest molecule rotation. For modeling the solid-state structure, up to three host-guest units are considered and the convergence with respect to the size of the solid-state fragment is investigated.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D76722
1,4-Dicyanobenzene, 98%
C8H4N2