EMAIL THIS PAGE TO A FRIEND
Behavioural pharmacology
Latent inhibition is attenuated by noise and partially restored by a 5-HT2A receptor antagonist.
PMID 12478218
Abstract
Latent inhibition (LI) is a model of attention, which is a cognitive process that can be modulated by stressors such as chronic intermittent broadband noise, e.g. caused by building work, which is particularly stressful to rats. The aim of this study was to analyse the effect of chronic noise stress, caused by a building project, on LI, and its interaction with SR 46,349B, a 5-HT2A receptor antagonist. Control groups from LI experiments conducted during periods of chronic intermittent noise were compared with control groups from LI experiments conducted in normal quiet conditions. The interaction of SR 46,349B with the effects of chronic noise stress was then tested. Chronic intermittent noise attenuated LI, an effect which was partially reversed by SR 46,349B, 2.4 mg/kg i.p. Attenuation of LI by chronic intermittent noise and reversal of this effect by SR 46,349B support suggestions that stress can modulate attention and that 5-HT2A receptors are involved in mediating the effects of chronic stress.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S7201
SR-46349 hemifumarate salt, ≥98% (HPLC)
C19H21FN2O2 · 0.5 C4H4O4