EMAIL THIS PAGE TO A FRIEND
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association
Implication of nitro group reduction in the mutagenic and chromosome damaging activities of 22 new 5-nitroisoquinolines by the Salmonella mutagenicity test and the cytokinesis-blocked micronucleus assay.
PMID 12480303
Abstract
The mutagenic (MUT) and chromosome-damaging (CHR) activities of 22 potential antimalarial drugs (5-nitroisoquinoline derivatives) were evaluated by the Salmonella test and the cytokinesis-blocked micronucleus assay (CBMN). The Salmonella mutagenicity test was performed with and without metabolic activation (S9 mix) in S. typhimurium strains TA100 and YG1042 (an overproducing nitroreductase and O-acetyltransferase TA100 strain). The CBMN was carried out on human lymphocytes without metabolic activation. Four concentrations were tested: 1, 10, 100 and 1000 ng/ml. MUT was expressed as minimal mutagenic concentrations (MMC, microM) and CHR was expressed as minimal chromosome-damaging concentrations (MCDC, nM) to compare both activities. All the 5-nitroisoquinoline compounds were mutagenic in TA100. MMC ranged from 0.1 to 52.9 microM in TA100. A statistically significant decrease in MMC was observed in YG1042 (8 x 10(-3) to 3.5 microM), implicating reduction of the nitro group. Modulation of MUT by S9 mix was not significant in TA100 and YG1042. CHR was detected in 13 products for at least one concentration. Among the chromosome-damaging compounds, the MCDC ranged from 2.9 x 10(-3) to 3.6 nM. No relationship was found between MUT and CHR, suggesting two distinct pathways of DNA damage.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
130222
5-Nitroisoquinoline, 98%
C9H6N2O2