EMAIL THIS PAGE TO A FRIEND
American journal of contact dermatitis : official journal of the American Contact Dermatitis Society
Iodopropynyl butylcarbamate as a preservative.
PMID 12484387
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
521949
3-Iodo-2-propynyl N-butylcarbamate, 97%
C8H12INO2