EMAIL THIS PAGE TO A FRIEND
Revue de pneumologie clinique
[Anti-Pseudomonas aerosol therapy in cystic fibrosis: improvement with tobramycin].
PMID 12486796
Abstract
Aerosol delivery of antibiotics offers the potential to achieve high antibiotic concentrations at the site of infection while reducing the risk of systemic untoward effects because of minimal resorption into the bloodstream. We reviewed knowledge acquired in this field over the two latter decades. While the earliest data were obtained with gentamycin, the most conclusive evidence presently regards aminoglycosides and colistin. Aerosol delivery of tobramycin was recently improved with the development of a new formulation for inhalation. Coupled with an adequate nebulization system, intermittent treatment with tobramycin for inhalation has been evaluated in randomized placebo-controlled studies. These studies have demonstrated a significant improvement of respiratory function.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Y0001210
Tobramycin for identification, European Pharmacopoeia (EP) Reference Standard
C18H37N5O9