EMAIL THIS PAGE TO A FRIEND
Chemistry (Weinheim an der Bergstrasse, Germany)
Generation and use of an equivalent of difluoroacetamide or difluoroacetate anions.
PMID 12489220
Abstract
Ethers of trifluoroacetaldehyde hemiaminals can undergo dehydrofluorination under basic conditions to provide ethers of difluoroketene hemiaminals. The latter behave as equivalents of difluoroacetamide or difluoroacetate anions towards various electrophiles, yielding a range of difluoromethylcarbonyl products.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
142859
Difluoroacetic acid, 98%
C2H2F2O2