EMAIL THIS PAGE TO A FRIEND
Methods in molecular biology (Clifton, N.J.)
Chemical modifications of proteins as an aid to sequence analysis.
PMID 12489442
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
06116
6-(Iodoacetamido)fluorescein, suitable for fluorescence, ≥95% (HPLC)
C22H14INO6