EMAIL THIS PAGE TO A FRIEND
The Journal of pharmacology and experimental therapeutics
Mitochondrial dysfunction is an early manifestation of 1,1-dichloroethylene-induced hepatotoxicity in mice.
PMID 12490582
Abstract
Hepatotoxicity induced by 1,1-dichloroethylene (DCE) is mediated by cytochrome P450-dependent metabolism to reactive intermediates, including the epoxide. We have tested the hypothesis that mitochondria are a primary target of toxicity by investigating dose- and time-dependent effects of DCE on mitochondrial respiration. Hepatotoxicity, as assessed by serum alanine aminotransferase (ALT) activity, was evaluated. We have also determined the effectiveness of N-acetyl-L-cysteine (NAC) in protecting against respiratory perturbations and hepatotoxicity. Liver mitochondria were isolated 2 h after DCE (50, 75, 100, 125, and 150 mg/kg) treatment. Glutamate (complex I)- and succinate (complex II)-supported mitochondrial respiration was assessed by measurement of state 3 (ADP-stimulated) and state 4 (resting) rates of oxygen consumption. The corresponding respiratory control ratios (RCRs, state 3/state 4) and ADP:O ratios were then calculated. A DCE dose of 125 mg/kg significantly inhibited glutamate- and succinate-supported state 3 respiration, leading to a significant reduction in corresponding RCRs and ADP:O ratios. In time-dependent studies, state 3 respiration rates and RCRs for glutamate-supported respiration were significantly decreased as early as 20 min after DCE (125 mg/kg) treatment, whereas those for succinate-supported respiration were significantly decreased at 90 min. Additionally, ADP:O ratios for glutamate-supported respiration were significantly decreased starting at 60 min, and those for succinate-supported respiration at 90 min. Alterations in mitochondrial function preceded significant increases in ALT activity, which was first manifested at 2 h. Pretreatment with NAC (1200 mg/kg) abrogated DCE-induced GSH depletion and inhibited disturbances in mitochondrial respiration. Moreover, NAC protected against increased ALT activity, suggesting that the protective effect of NAC is due to increased GSH for conjugation reactions and/or its antioxidant property. These results showed that DCE-mediated mitochondrial dysfunction is an early event that preceded the onset of hepatotoxicity.