EMAIL THIS PAGE TO A FRIEND
Contact dermatitis
Allergic contact dermatitis from trimethylolpropane triacrylate and pentaerythritol triacrylate.
PMID 12492540
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
246794
Pentaerythritol triacrylate, technical grade
C14H18O7