EMAIL THIS PAGE TO A FRIEND
The Journal of Biological Chemistry
A Kunitz-type protease inhibitor, bikunin, inhibits ovarian cancer cell invasion by blocking the calcium-dependent transforming growth factor-beta 1 signaling cascade
PMID 12496270
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
05-168
Anti-TNFα Antibody, clone MPG-XT3, clone MPG-XT3, Upstate®, from rat