EMAIL THIS PAGE TO A FRIEND
Adverse drug reactions and toxicological reviews
Adverse drug reaction update.
PMID 12503258
Abstract
Increasing numbers of articles on adverse drug reactions are published in a wide range of medical journals. To help keep you up-to-date with the latest advances worldwide on all aspects of adverse drug reactions, this section of the journal brings you information selected from the drug safety alerting service Reactions Weekly. The following reports are selected from the latest issues, summarizing the most important clinical studies, adverse reaction news, and expert opinion pieces published across a broad range of literature sources.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
59780
DL-Kavain, analytical standard
C14H14O3