EMAIL THIS PAGE TO A FRIEND
The Annals of pharmacotherapy
Niacin-ER and lovastatin treatment of hypercholesterolemia and mixed dyslipidemia.
PMID 12503944
Abstract
To review the currently available information on the once-daily combination of niacin extended-release (ER)/lovastatin in the treatment of patients with hypercholesterolemia and mixed dyslipidemia at high risk for cardiovascular events. MEDLINE (1966-July 2002) was searched for primary and review articles. Data from the manufacturer were also included. All articles and product labeling deemed relevant to the combination of niacin and statins (i.e., lovastatin) were included for review. English-language studies selected for inclusion were limited to those with human subjects. The Food and Drug Administration approved a new fixed-dose combination of niacin-ER/lovastatin, which is administered once daily. The efficacy and safety of the combined agent have been proven to be similar to either component used alone or in combination for management of hyperlipidemia and mixed dyslipidemia. Elevated low-density lipoprotein cholesterol (LDL-C) is independently associated with a higher risk for cardiovascular events. Lowering of elevated LDL-C concentrations with statin monotherapy may be insufficient in patients at high risk for cardiovascular events. In fact, consideration of elevated triglycerides (TGs) and/or low concentrations of high-density lipoprotein cholesterol (HDL-C) in patients with elevated LDL-C places them at greater risk. The addition of niacin may enhance or improve the lipid profile of those who require a further decrease of TGs and/or increase of HDL-C even after stable statin therapy. Niacin-ER offers efficacy similar to that of immediate-release niacin, but minimal myopathy and hepatotoxicity (compared with sustained-release niacin). Although no clinical outcomes are available, current evidence shows that the combination product offers adequate lowering of LDL-C and TGs and increasing HDL-C. The data suggest that therapy with the niacin-ER and lovastatin combination product is safe and does not increase the incidence of adverse effects.