EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences
Determination of 6-hydroxychlorzoxazone and chlorzoxazone in porcine microsome samples.
PMID 12504188
Abstract
A simple, accurate and sensitive HPLC method for the in vitro determination of 6-hydroxychlorzoxazone and chlorzoxazone in porcine microsome samples is described. Chromatography was performed on a YMC-Pack ODS-AQ column using a mobile phase of 0.05% phosphoric acid pH 3-methanol (60:40, v/v). UV detection was carried out at 287 nm. The detector response was linear over the concentration range 25-2000 ng/ml. This assay produced quick, accurate, and repeatable results.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
UC148
6-Hydroxychlorzoxazone, ≥98% (HPLC)
C7H4ClNO3
C188
6-Hydroxychlorzoxazone, solid
C7H4ClNO3